KidWind® Motor Mount Assembly
KidWind motor mount assembly
Catalog # CA470165-148
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: KW-MTRMNT
KidWind® Motor Mount Assembly
Catalog # CA470165-148
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
The KidWind Motor Mount Pack holds the motor (generator) in place when affixed to the wind turbine nacelle. The Motor Mount Pack can be used to replace lost parts from a nacelle. It can also be used to mount a nacelle to the tower for a vertical axis wind turbine configuration.
Spécifications
- Désignation:KidWind motor mount assembly