Colored Beads
Colored Beads, 13 mm
Catalog # 470089-446
Supplier: PRECISION MOULDED POLYMERS LTD
Fournisseur #: AM1090.00000.00000
Jump to:
Product Details & Documents
Sorted colors. Plastic beads have 3.5 mm hole and 13 mm diameter. Set of 216.
These assorted spherical beads are made of brightly colored plastic in red, orange, yellow, green, blue and purple. The colors are evenly distributed in each pack, 21 of each color. Each beads is 13 mm in diameter with a 3.5 mm hole, excellent for sorting, matching, and threading or any other craft needs. Comes in a pack of 126 beads.
Spécifications
- Ø:13 mm
- Désignation:Colored Beads