ContactUsLinkComponent
VWR® Freezer Verification Thermometer with Certificate of Compliance, Traceable to NIST
VWR® Freezer Verification Thermometer with Certificate of Compliance, Traceable to NIST
Catalog # 89495-970
Supplier:  VWR International
undefined
VWR® Freezer Verification Thermometer with Certificate of Compliance, Traceable to NIST
Catalog # 89495-970
Supplier:  VWR International
Specifications
  • Length:
    135 mm (5.3”)
  • Cat. no.:
    89495-970
  • Divisions:
    0.5 °C
  • Accuracy:
    ±0.5 °C
  • Range:
    −30…+1 °C
  • Certification Points:
    -15°C
Specifications
About this item
These thermometers are certified at one specific temperature.
  • PFA coating
  • Mercury-free
  • Permanently fused markings
  • Lead-free glass
PFA coated for enhanced safety in case of breakage, they are used to verify sample temperatures in freezers covering ranges from -30 to 1°C
Ideal for the chemical, petro-chemical, pharmaceutical, medical, cosmetic, and dairy industries, these thermometers are found in universities, breweries, refrigeration houses, scientific institutes, blood banks, freezer and laboratory equipment producing facilities.
These mercury-free thermometers are filled with EnviroKleen™ certified Enviro-Safe® liquid, and have permanently fused markings and lead-free glass. Insertion of the thermometer into the included recyclable plastic bottle, filled with glass beads or EnviroKleen™ certified Enviro-Safe® liquid, protects samples from contamination by eliminating the need for direct contact. Bottle contents mimic the sample for more accurate temperature monitoring. Enviro-Safe® thermometer fill/Enviro-Safe® bottle fill.
Certifications: Manufactured and tested by Avantor's USA quality team, these verification thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Individually serialized.