Elma Clean 260 Dip and Splash Gentle Ultrasonic Cleaning Solution, Elma
Supplier: Elma
Certificates
About this item
Neutral, biodegradable Elma clean 260 dip and splash gently cleans sensitive surfaces. The cleaning solution is easily rinsed off with water leaving no residue. Use in an ultrasonic cleaner or labware washer
- Ultrasonic cleaning concentrate for sensitive metals, plastics, and glass
- Removes coolant, grease, oil, dust, fingerprints, and lime soaps
- Ideal for cleaning aluminum prior to anodizing or coating
- Contains no silicates and rinses residue-free
Elma clean 260 dip and splash is highly concentrated and should be diluted to 1-2% with water before use. A 10 liter bottle dilutes to 132 gallons of 2% solution.
Oil and grease removed from parts will separate and float on the liquid surface where they can be skimmed off.
Safe and cost effective Elma cleaning solutions prolong the lifetime of your ultrasonic bath.
Specifications
- Catalog No:
- 10724-602
- 10724-604
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health