tec clean A1 Ultrasonic Cleaning Solution for Electronics, Optics, and Plastics, Elma
Fournisseur: Elma
Certificats
À propos de cet article
Biodegradable Elma tec clean A1 is highly concentrated and should be diluted to 3-10% with water before use
Safe and cost effective Elma cleaning solutions prolong the lifetime of your ultrasonic bath.
A 10 liter bottle dilutes to 53 gallons of 5% solution.
Spécifications
- Catalog No:
- 10724-594
- 10724-596
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health