tec clean S1 Ultrasonic cleaning Solution for Corrosion Removal, Elma
Catalog # 10724-590
Supplier: Elma
Specifications
- Size:10 L
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Cat. no.:10724-590
- Supplier No.:8000161
Specifications
About this item
Safe and cost effective Elma cleaning solutions prolong the lifetime of your ultrasonic bath.
- Elma tec clean S1
- Mildly acidic cleaning concentrate
- Removes oxide film, rust, lime, grease, oil, corrosion, and mineral deposits
- Rinses residue-free
- Safely removes oxidation and corrosion from electronic parts and printed circuit boards
- Brightens brass and copper alloys
Biodegradable Elma tec clean S1 is highly concentrated and should be diluted to 1-5% with water before use. A 10 liter bottle dilutes to 132 gallons of 2% solution.