Student gratings
Student gratings
Student gratings
Student gratings
Quality gratings perfect for your Optics Needs
  • Viewing area measures approx. 45x30mm
Quality transmission gratings protected by two glass plates, 60x45mm.
Product Family Options
  • Désignation
  • Student gratings, 300 lines per mm
    Each
    Each - $85.64
    $85.64
    Each
       $85.64
     : DFG300
  • Student gratings, 100 lines per mm
    Each
    Each - $85.65
    $85.65
    Each
       $85.65
     : DFG100
  • Student gratings, 600 lines per mm
    Each
    Each - $85.65
    $85.65
    Each
       $85.65
     : DFG600