Easy 50R Ultrasonic Sieve Cleaner
Catalog # 10724-488
Supplier: Elma
CSA Certified
UL Listed
Specifications
- Frequency:37 kHz (ultrasonic)
- Capacity:1.4 gal.
- Description:Easy 50R Ultrasonic Sieve Cleaner
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Electrical:110-120 VAC, 50/60Hz
- Tank dimensions Ø×H:9.4 x 5.0"
- Timer:1-30 minutes, continuous
- Cat. no.:10724-488
- Supplier No.:1049548
Specifications
About this item
The powerful Elmasonic Easy 50R ultrasonic cleaner has 3 operating modes optimized for cleaning of sieves and lab instruments, degassing of solvents, and preparation of laboratory samples.
- Ultrasonic bath for all standard laboratory applications
- 3 optimized modes: Sieve cleaning, Degassing, Sample preparation
- Intuitive control panel
- High intensity ultrasonic power
- Cylindrical tank designed for 8-inch sieves and 1-liter solvent bottles
Dependable, state-of-the-art Elma ultrasonic cleaners have a timer which can be set from 1-30 minutes or continuous operation. For safety purposes the unit turns off automatically after 12 hours of continuous operation. The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run.
Certifications: CE, UL, CSA, ISO9001, ISO13485.
Ordering information: The cover and basket are available and are ordered separately.