VWR® Precision Scissors
Precision scissors, Style No. SS, 140 mm, Stainless steel
Catalog # 89259-974
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Precision Scissors are stainless steel with very sharp, fine points.
- Anti-magnetic properties
- Resistant to acids
- Scissors feature fine points
Precision scissors are available in a range of lengths and order sizes, from individual to bulk.
Spécifications
- Désignation:Precision scissors, Style No. SS
- Longueur:140 mm
- Matière:Stainless steel