Low Ash Quantitative Paper Grade 388, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 388 is a fast filtering low ash (Ash 0.01%) filter paper suitable for routine quantitative gravimetric techniques.
It is used for rapid filtration of coarse particles.
Specifications
- Catalog No:
- 10754-432
- 10754-430
- 10754-436
- 10754-502
- 10754-434
- 10754-500
- 10754-506
- 10754-438
- 10754-504
- 10754-422
- 10754-426
- 10754-424
- 10754-428
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns
- 12-15 microns