Anti-IgG Donkey Antibody (CF™ 633)
Catalog # 89493-230
Supplier: Biotium
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:CF® 633
- ELISA:Yes
- Flow cytometry:Yes
- Host:Donkey
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Size:1 mg
- Cat. no.:89493-230
- Supplier No.:20124-1MG
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 633
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse