Anti-IgG Mouse Monoclonal Antibody (CF™ 594)
Catalogue # 89411-294
Fournisseur: Biotium
Spécifications
- Antibody type:Primary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:CF® 594
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Western blot:Yes
- Size:50 µL
- Cat. no.:89411-294
- Supplier No.:20205-1
Spécifications
À propos de cet article
Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: CF™ 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Biotin