Anti-PRKAR2A Mouse Monoclonal Antibody [clone: 1B4]
Catalog # 89272-454
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase cAMP-dependent Regulatory Type 2 Alpha
- Antigen symbol:PRKAR2A
- Clonality:Monoclonal
- Clone:1B4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:5576
- Antigen synonyms:PKR2|PRKAR2
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PRKAR2A (NP_004148) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. no.:89272-454
- Supplier No.:GTX83826
Specifications
About this item
Mouse monoclonal antibody [1B4] to PRKAR2A
Recommended Starting Dilutions:WB 1:1000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PRKAR2A
Clonality: Monoclonal
Clone: 1B4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human