Anti-ADCY6 Chicken Polyclonal Antibody
Catalogue # 89263-748
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Adenylate Cyclase 6
- Antigen symbol:ADCY6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Size:50 μg
- Gene ID:112
- Antigen synonyms:LCCS8|KIAA0422|AC6
- Storage buffer:Phosphate-Buffered Saline containing no preservatives
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide - human Adenylate cyclase type 6
- Purification:Affinity purified with antigen
- Cat. no.:89263-748
- Supplier No.:GTX82042
Spécifications
À propos de cet article
Chicken polyclonal to Adenylate cyclase type 6
WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.
Type: Primary
Antigen: ADCY6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human