Anti-CGA Rabbit Monoclonal Antibody [clone: SP12]
89285-552
: Genetex
- Antibody type:Primary
- Antigen name:Chromogranin A
- Antigen symbol:CgA
- Clonality:Monoclonal
- Clone:SP12
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:500 μl
- Gene ID:1113
- Antigen synonyms:betagranin (N-terminal fragment of chromogranin A)|SP-I|chromogranin-A|pituitary secretory protein I|parathyroid secretory protein 1
- Storage buffer:This antibody is supplied as tissue culture supernatant containing sodium azide as preservative.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Recombinant protein encoding human Chromogranin A.
- Cat. no.:89285-552
- Supplier No.:GTX79418
Monoclonal Rabbit Antibody to Human Chromogranin A
We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: SP12
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: