Anti-CD10 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT10]
Catalogue # 89285-600
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Neprilysin
- Antigen symbol:CD10
- Clonality:Monoclonal
- Clone:LT10
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:4311
- Antigen synonyms:Calla|Common Acute Lymphoblastic Leukemia Antigen|Nep|Mme|Cd10|Membrane Metallo-Endopeptidase|Cluster Of Differentiation 10|Neutral Endopeptidase
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:mouse NALM-6 leukemia pre-B cell line (tissue/cell preparation)
- Cat. no.:89285-600
- Supplier No.:GTX78376
Spécifications
À propos de cet article
Monoclonal Antibody to CD10 (Human) Fluorescein (FITC)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: LT10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human