Anti-IL2RA Mouse Monoclonal Antibody (Biotin) [clone: MEM-140]
89285-524
: Genetex
- Antibody type:Primary
- Antigen name:Interleukin 2 Receptor, Alpha
- Antigen symbol:IL2RA
- Clonality:Monoclonal
- Clone:MEM-140
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Size:100 μg
- Gene ID:3559
- Antigen synonyms:TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- Storage buffer:Phosphate buffered saline (PBS) with 15 mM sodium azide, approx. pH 7.4
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:PHA-activated peripheral blood leucocytes.
- Cat. no.:89285-524
- Supplier No.:GTX78341
Monoclonal Antibody to CD25 (Human) Biotin
The reagent is designed for Flow Cytometry analysis.Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: MEM-140
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human