To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neprilysin
- Antigen symbol:CD10
- Clonality:Monoclonal
- Clone:MEM-78
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:4311
- Antigen synonyms:Calla|Common Acute Lymphoblastic Leukemia Antigen|Nep|Mme|Cd10|Membrane Metallo-Endopeptidase|Cluster Of Differentiation 10|Neutral Endopeptidase
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Pre-B cell line NALM-6.
- Cat. no.:89285-570
- Supplier No.:GTX78263
Specifications
About this item
Monoclonal Antibody to CD10 (Human) Fluorescein (FITC)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 _l reagent / 100 _l of whole blood or 106 cells in a suspension. The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: MEM-78
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human