Anti-TBX19 Rabbit Polyclonal Antibody
Catalog # 89286-692
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:T-box 19
- Antigen symbol:TBX19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Gene ID:9095
- Antigen synonyms:TPIT|TBS19|dJ747L4.1
- Storage buffer:PBS with 2% sucrose
- Storage temperature:Store at -20 °C or below. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human TBX19
- Purification:IgG fraction
- Cat. no.:89286-692
- Supplier No.:GTX77878
Specifications
About this item
Rabbit Polyclonal Antibody to TBX19
WB: Suggested dilution at 1.0-1.5 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: TBX19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human