Anti-TOP2A Rabbit Polyclonal Antibody
Catalogue # 89286-680
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Topoisomerase 2 alpha
- Antigen symbol:TOP2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Gene ID:7153
- Antigen synonyms:alpha isozyme|DNA topoisomerase 2-alpha|DNA gyrase|DNA topoisomerase (ATP-hydrolyzing)|TP2A|170 kD|DNA topoisomerase II|TOP2
- Storage buffer:PBS with 2% sucrose
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human TOP2A
- Purification:Protein A/G affinity purified
- Cat. no.:89286-680
- Supplier No.:GTX77787
Spécifications
À propos de cet article
Rabbit Polyclonal Antibody to Topoisomerase Iia
WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: TOP2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human