Anti-NFKBIA Rabbit Polyclonal Antibody
Catalogue # 89286-662
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Nuclear Factor of Kappa Light Polypeptide Gene Enhancer in B-cells Inhibitor, Alpha
- Antigen symbol:NFKBIA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Gene ID:4792
- Antigen synonyms:MAD3|IkappaBalpha|IkB-alpha|Major histocompatibility complex enhancer-binding protein MAD3|NFKBI|NF-kappa-B inhibitor alpha|I-kappa-B-alpha|IKBA
- Storage buffer:PBS with 2% sucrose
- Storage temperature:Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NFKBIA
- Purification:Protein A/G affinity purified
- Cat. no.:89286-662
- Supplier No.:GTX77737
Spécifications
À propos de cet article
Rabbit Polyclonal Antibody to NFKBIA
WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human