Anti-CD62L Mouse Monoclonal Antibody [clone: OX-85]
Catalog # 89285-210
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:L-Selectin
- Antigen symbol:CD62L
- Clonality:Monoclonal
- Clone:OX-85
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:250 μg
- Gene ID:29259
- Antigen synonyms:LECAM1|SELL|CD62L|TQ1|LEU8|LNHR|LYAM1vPLNHR|LSEL|LAM1
- Storage buffer:PBS, pH7.2 containing 0.02% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant rat L-selectin
- Purification:Protein G affinity purified
- Cat. no.:89285-210
- Supplier No.:GTX74988
Specifications
About this item
Mouse monoclonal [OX-85] to CD62L
For ELISA: Use at an assay dependent dilution. For FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: OX-85
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: