Anti-Cl channel interacting Protein (SID1 transmembrane Family member 1) Rabbit Polyclonal Antibody
Catalogue # 89275-854
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Sid1 Transmembrane Family, Member 1
- Antigen symbol:Cl channel interacting Protein (SID1 transmembrane Family member 1)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:50 μg
- Gene ID:54847
- Antigen synonyms:B830021E24Rik|SID-1|SID1
- Storage buffer:Phosphate, pH7.2-7.4
- Storage temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. no.:89275-854
- Supplier No.:GTX47826
Spécifications
À propos de cet article
FITC-conjugated Rabbit polyclonal antibody to Cl channel interacting protein
Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: Cl channel interacting Protein (SID1 transmembrane Family member 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: