Anti-CD317 Rabbit Polyclonal Antibody
89275-832
: Genetex
- Antibody type:Primary
- Antigen name:Bone Marrow Stromal Cell Antigen 2
- Antigen symbol:CD317
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 μg
- Gene ID:684
- Antigen synonyms:Tetherin|BST2|PDCA-1
- Storage buffer:Phosphate, pH7.2-7.4
- Storage temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. no.:89275-832
- Supplier No.:GTX47801
FITC-conjugated Rabbit polyclonal antibody to CD317
We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: CD317
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: