Anti-UBLCP1 Rabbit Polyclonal Antibody
89267-844
: Genetex
- Antibody type:Primary
- Antigen name:Ubiquitin-like Domain Containing CTD phosphatase 1
- Antigen symbol:UBLCP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 μg
- Gene ID:134510
- Antigen synonyms:CPUB1
- Storage buffer:Phosphate buffered saline (PBS)
- Storage temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. no.:89267-844
- Supplier No.:GTX45699
Rabbit polyclonal antibody to UBLCP1 (N-terminal)
Recommended Dilutions: For Western Blot: Use at 1 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: UBLCP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat