Anti-SPATA5 Rabbit Polyclonal Antibody
89267-836
: Genetex
- Antibody type:Primary
- Antigen name:Spermatogenesis Associated 5
- Antigen symbol:SPATA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 μg
- Gene ID:166378
- Antigen synonyms:AFG2|EHLMRS|SPAF
- Storage buffer:Phosphate buffered saline (PBS)
- Storage temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. no.:89267-836
- Supplier No.:GTX45695
Rabbit polyclonal antibody to SPATA5 (Middle)
Recommended Dilutions: For Western Blot: Use at 1 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: SPATA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat