Anti-CNTD1 Rabbit Polyclonal Antibody
Catalog # 89267-614
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Cyclin N-terminal Domain Containing 1
- Antigen symbol:CNTD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 μg
- Gene ID:124817
- Antigen synonyms:CNTD
- Storage buffer:Phosphate buffered saline (PBS)
- Storage temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. no.:89267-614
- Supplier No.:GTX45580
Specifications
About this item
Rabbit polyclonal antibody to CNTD1 (N-terminal)
Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CNTD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: