Anti-C1orf122 (chromosome 1 open reading frame 122) Rabbit Polyclonal Antibody
89267-500
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Chromosome 1 Open Reading Frame 122
- Antigen symbol:C1orf122 (chromosome 1 open reading frame 122)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 μg
- Gene ID:127687
- Antigen synonyms:ALAESM
- Storage buffer:Phosphate buffered saline (PBS)
- Storage temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. no.:89267-500
- Supplier No.:GTX45522
Rabbit polyclonal antibody to C1orf122 (N-terminal)
Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: C1orf122 (chromosome 1 open reading frame 122)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human