Order Entry
Canada
ContactUsLinkComponent
Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 39266]
Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 39266]
  89288-316
 :  Genetex
Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 39266]
  89288-316
 :  Genetex
 :  GTX43419

 

  • Antibody type:
    Primary
  • Antigen name:
    Ly-6B.2 AlloAntigen
  • Antigen symbol:
    Ly-6B.2 AlloAntigen
  • Clonality:
    Monoclonal
  • Clone:
    39266
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Reactivity:
    Mouse
  • Size:
    100 µg
  • Cat. no.:
    89288-316
  • Supplier No.:
    GTX43419

 

 

Type: Primary
Antigen: Ly-6B.2 AlloAntigen
Clonality: Monoclonal
Clone: 39266
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse