Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
Catalog # 89288-286
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:HCN1
- Antigen symbol:HCN1
- Clonality:Monoclonal
- Clone:S70-28
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μg
- Gene ID:84390
- Storage buffer:PBS pH7.4, 50% glycerol
- Storage temperature:Aliquot and store at -20 C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 778-910 (C terminus) of rat HCN1, accession number Q9JKBO
- Purification:Protein G Purified
- Cat. no.:89288-286
- Supplier No.:GTX41985
Specifications
About this item
Mouse monoclonal antibody [S70-28] to HCN1
We recommend a working dilution of 1-10ug/mL (WB), 0.1-1.0ug/mL (Perox) (IHC/ICC), 1.0-10ug/mL (IF).
Type: Primary
Antigen: Hcn1
Clonality: Monoclonal
Clone: S70-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat