Anti-DECTIN-1 Rat Monoclonal Antibody [clone: 2A11]
89333-502
: Genetex
- Antibody type:Primary
- Antigen name:C-type lectin Domain Family 7 member A
- Antigen symbol:DECTIN-1
- Clonality:Monoclonal
- Clone:2A11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 μg
- Gene ID:56644
- Antigen synonyms:DC-associated C-type lectin 1|BGR|Clecsf12|beta-glucan receptor|dendritic cell-associated C-type lectin 1|carbohydrate recognition domain) lectin|dectin-1|beta-GR,C-type (calcium dependent
- Storage buffer:Phosphate buffered saline
- Storage temperature:Store at +4°C. DO NOT FREEZE. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Dectin-1 transfected NIH3T3 cells and recombinant soluble Dectin-1
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89333-502
- Supplier No.:GTX41471
Rat monoclonal antibody [2A11] to Dectin-1
Recommended Starting Dilutions:Flow Cytometry: 1/10 - 1/50Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: DECTIN-1
Clonality: Monoclonal
Clone: 2A11
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse