Anti-Escherichia coli J5 LPS Mouse Monoclonal Antibody [clone: 2D7/1]
Catalog # 89285-646
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:E. coli J5 LPS
- Antigen symbol:Escherichia coli J5 LPS
- Clonality:Monoclonal
- Clone:2D7/1
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:E. coli
- Size:100 µg
- Cat. no.:89285-646
- Supplier No.:GTX41231
Specifications
About this item
Type: Primary
Antigen: Escherichia coli J5 LPS
Clonality: Monoclonal
Clone: 2D7/1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: E. coli Bacteria