Order Entry
Canada
ContactUsLinkComponent
Anti-IgA Rat Monoclonal Antibody [clone: LO-MA-10]
Anti-IgA Rat Monoclonal Antibody [clone: LO-MA-10]
  89285-634
 :  Genetex
Anti-IgA Rat Monoclonal Antibody [clone: LO-MA-10]
  89285-634
 :  Genetex
 :  GTX40467

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin A
  • Antigen symbol:
    IgA
  • Clonality:
    Monoclonal
  • Clone:
    LO-MA-10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rat
  • Isotype:
    IgM
  • Reactivity:
    Mouse
  • Size:
    250 μg
  • Epitope:
    Heavy chain
  • Gene ID:
    16061
  • Storage buffer:
    Phosphate buffered saline, 0.09% Sodium Azide (NaN3)
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Cat. no.:
    89285-634
  • Supplier No.:
    GTX40467

 

 

Rat anti-Mouse IgA (Heavy chain) Monoclonal antibody [LO-MA-10]

Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: LO-MA-10
Conjugation:
Epitope: Heavy chain
Host: Rat
Isotype:
Reactivity: Mouse