Order Entry
Canada
ContactUsLinkComponent
 
Anti-IFNG Mouse Monoclonal Antibody [clone: CC302]
  89333-496
 :  Genetex
Anti-IFNG Mouse Monoclonal Antibody [clone: CC302]
  89333-496
 :  Genetex
 :  GTX39976

 

  • Antibody type:
    Primary
  • Antigen name:
    Interferon Gamma
  • Antigen symbol:
    IFNG
  • Clonality:
    Monoclonal
  • Clone:
    CC302
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Horse,
    Mustelid,
    Sheep,
    Dolphin,
    Goat,
    Human,
    Bovine,
    Dog
  • Size:
    100 μg
  • Gene ID:
    281237
  • Storage buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide (NaN3)
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. no.:
    89333-496
  • Supplier No.:
    GTX39976

 

 

Mouse monoclonal antibody [CC302] to IFN gamma

Recommended Starting Dilutions:Flow Cytometry: 1/100 - 1/500Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: CC302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Pig, Dolphin, Ferret, Goat, Horse, Mink, Sheep, Bovine