Anti-STAT4 Rabbit Polyclonal Antibody
89278-666
: Genetex
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 4
- Antigen symbol:STAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:1 ml
- Gene ID:20849
- Antigen synonyms:SOCS6|STATI4|SLEB11|STAI4|SOCS4|HSPC060|Signal Transducer and Activator of Transcription|CIS4|SSI4
- Storage buffer:PBS with 0.1% sodium azide
- Storage temperature:Store this product at 4 °C, do not freeze. The product is stable for one year from the date of shipment.
- Concentration:0.2 mg/ml
- Immunogen:Synthetic peptide (Mouse) (C terminal).
- Purification:IgG fraction
- Cat. no.:89278-666
- Supplier No.:GTX27967
Rabbit polyclonal to STAT4
Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat