Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 89285-830
Supplier: Genetex
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Sheep
- Western blot:Yes
- Size:1 mg
- Cat. no.:89285-830
- Supplier No.:GTX26748
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep