Anti-RGS1 Goat Polyclonal Antibody
89286-300
: Genetex
- Antibody type:Primary
- Antigen name:Regulator of G-protein Signaling 1
- Antigen symbol:RGS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 μg
- Gene ID:5996
- Antigen synonyms:B-cell activation protein BL34|BL34|IER1|regulator of G-protein signalling 1|1R20|immediate-early response 1|early response protein 1R20|IR20|Regulator of G-protein signaling 1|B-cell specific
- Storage buffer:Tris saline, 0.02% Na azide, pH7.3 with 0.5% BSA
- Storage temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence NLLNDLQANSLK, from C Terminus of the protein sequence according to NP_002913
- Purification:IgG fraction
- Cat. no.:89286-300
- Supplier No.:GTX22715
Western Blot - Approx 25kDa band seen in HepG2 lysate. Recommended for use at 0.5-2.0?g/ml.
Type: Primary
Antigen: RGS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse