Order Entry
Canada
ContactUsLinkComponent
Anti-STAT6 Rabbit Polyclonal Antibody
Catalogue # 89278-644
Fournisseur:  Genetex
Anti-STAT6 Rabbit Polyclonal Antibody
Catalogue # 89278-644
Fournisseur:  Genetex
Supplier Number:  GTX22424

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Signal Transducer And Activator of Transcription 6
  • Antigen symbol:
    STAT6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    1 ml
  • Gene ID:
    6778
  • Antigen synonyms:
    STAT6 pY641|STAT6C|IL-4-STAT|interleukin 4 induced STAT|D12S1644|STAT6B
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: 0.2% Gelatin, PBS
  • Storage temperature:
    Store this product at 4 °C, do not freeze. The product is stable for one year from the date of shipment.
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    Synthetic peptide (Human) (C terminal).
  • Cat. no.:
    89278-644
  • Supplier No.:
    GTX22424

Spécifications

À propos de cet article

For Western Blotting, use at a dilution of 1:100 Optimal dilutions should be determined by the user.

Type: Primary
Antigen: STAT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human