Anti-SCP3 Rabbit Polyclonal Antibody
Catalogue # 89264-698
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Synaptonemal Complex Protein 3
- Antigen symbol:SCP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:50511
- Antigen synonyms:Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: Tris citrate/phosphate. pH 7-8
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (Human) (C terminal).
- Purification:Immunogen affinity purified
- Cat. no.:89264-698
- Supplier No.:GTX15093
Spécifications
À propos de cet article
IF: Use at a dilution of 1/100. IHC-P: Use at a dilution of 1/400. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: SCP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse