Anti-NTR2 Rabbit Polyclonal Antibody
Catalogue # 89277-692
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Neurotensin Receptor 2
- Antigen symbol:NTR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25μg
- Gene ID:23620
- Antigen synonyms:NTR2
- Storage buffer:Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (human; unfortunately, the amino acid sequence is considered to be commercially sensitive)
- Purification:Immunogen affinity purified
- Cat. no.:89277-692
- Supplier No.:GTX13068
Spécifications
À propos de cet article
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NTR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human