Anti-GPCR GPRC5B (G Protein-coupled Receptor Family C group 5 member B) Rabbit Polyclonal Antibody
Catalogue # 89277-644
Fournisseur: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor, Class C, Group 5, Member B
- Antigen symbol:GPRC5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25μg
- Gene ID:51704
- Antigen synonyms:RAIG2|RAIG-2
- Storage buffer:Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
- Purification:Immunogen affinity purified
- Cat. no.:89277-644
- Supplier No.:GTX12711
Spécifications
À propos de cet article
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GPRC5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human