Anti-EMAP II (aminoacyl tRNA Synthetase Complex-interacting multifunctional Protein 1) Goat Polyclonal Antibody
89286-082
: Genetex
- Antibody type:Primary
- Antigen name:Aminoacyl Trna Synthetase Complex-Interacting Multifunctional Protein 1
- Antigen symbol:EMAP II (aminoacyl tRNA Synthetase Complex-interacting multifunctional Protein 1)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 μg
- Gene ID:9255
- Antigen synonyms:HLD3|p43|EMAPII|SCYE1|EMAP2
- Storage buffer:Preservative: NoneConstituents: 0.5x PBS. pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Purification:Immunogen affinity purified
- Cat. no.:89286-082
- Supplier No.:GTX12442
ELISA: Use at a concentration of 0.5 ?g/ml. Sensitivity = 0.2-0.4 ng/well. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 37.4 kDa. Sensitivity = 1.5-3.0 ng/lane. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: EMAP II (aminoacyl tRNA Synthetase Complex-interacting multifunctional Protein 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human