Anti-LRRC23 Rabbit Polyclonal Antibody
100 μl
Catalog # 89324-422
Supplier: Genetex
Fournisseur #: GTX121536
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: LRRC23
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Spécifications
- Taille:100 μl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified by antigen-affinity chromatography.
- Antibody type:Primary
- Isotype:IgG
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Host:Rabbit
- Concentration:1 mg/ml
- Antigen name:Leucine Rich Repeat Containing 23
- Cat. No.:89324-422
- Reactivity:Human
- Gene ID:10233
- Antigen synonyms:LRPB7
- Immunogen:Recombinant protein encompassing a sequence within the center region of human LRRC23. The exact sequence is proprietary.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:LRRC23
- Storage buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.



