Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-OR8J1 Rabbit Polyclonal Antibody | Avantor

Anti-OR8J1 Rabbit Polyclonal Antibody

100 μl
Catalog # 89320-374
Supplier:   Genetex
Fournisseur #:  GTX109616
Anti-OR8J1 Rabbit Polyclonal Antibody
Catalog # 89320-374
Supplier:  Genetex
Anti-OR8J1 Rabbit Polyclonal Antibody

100 μl

Price
$949.51
Log in pour la disponibilité
Each (100µl)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Rabbit Polyclonal antibody to IMPA1 (inositol(myo)-1(or 4)-monophosphatase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR8J1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Spécifications
  • Taille:
    100 μl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified by antigen-affinity chromatography.
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Host:
    Rabbit
  • Concentration:
    1mg/ml
  • Antigen name:
    Olfactory Receptor, Family 8, Subfamily J Member 1
  • Cat. No.:
    89320-374
  • Reactivity:
    Human
  • Gene ID:
    3612
  • Antigen synonyms:
    IMPA|IMP
  • Immunogen:
    Full length human IMPA1 Recombinant protein.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    OR8J1
  • Storage buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.