ContactUsLinkComponent
HisTag® Antibody HRP Conjugate Kit, MilliporeSigma
Catalog # CA80017-612
Supplier:  MilliporeSigma
CAS Number:  
HisTag® Antibody HRP Conjugate Kit, MilliporeSigma
Catalog # CA80017-612
Supplier:  MilliporeSigma
Supplier Number:  71840-3
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    HisTag ® Antibody HRP Conjugate Kit
  • Size:
    125 ul
  • Storage temperature:
    2 to 8 °C
  • Cat. no.:
    CA80017-612
  • Supplier No.:
    71840-3
Specifications
About this item
The HisTag® Antibody HRP Conjugate kit contains a horseradish peroxidase-conjugated HisTag® Monoclonal Antibody that is useful as a single detection reagent, thus eliminating cross-reactivity associated with secondary antibody reagents
Using standard HRP detection methods, as little as 1 ng protein can be detected on dot blots. Cross-reactivity to bacterial, insect, yeast or mammalian cell proteins is negligible.
It recognises five consecutive histidine residues regardless of the surrounding amino acid context. The purified antibody conjugate produces a strong signal at a 1:1000 to 1:2000 dilution on Western blots and dot blots. The HisTag® Antibody HRP Conjugate also detects the recombinant marker bands in the Novagen Perfect Protein™ Western Markers for convenient visualisation of accurate internal standards on Western blots.
Ordering information: The package size provides enough conjugate for 25-50 blots and includes 5% Alkali-soluble Casein for the blocking reagent.