Neulog Chemistry Bundle | Avantor
Neulog Chemistry Bundle
NeuLog chemistry bundle
Catalog # 470117-616
Supplier:   Avantor
Neulog Chemistry Bundle
Catalog # 470117-616
Supplier:  Avantor
Neulog Chemistry Bundle
Neulog Chemistry Bundle
NeuLog chemistry bundle
Price
$1,084.85
Log in pour la disponibilité
Pack of 1
Product Details & Documents
A set of five NeuLog sensors that are great for use in chemistry classrooms.
  • Collect data in the classroom or remotely
  • Compatible with PC, Mac, and chromebook
  • Free software available for download
  • Connect multiple sensors simultaneously to a single battery or connectivity module
The NeuLog probeware system is a complete data collection platform perfect for classroom use. Data are sent from the sensors to any computer via USB, bluetooth, or WiFi. A rechargeable battery makes remote data collection easy: the sensors will store the data for pre-defined experiments until they are uploaded to a computer later. The free software is powerful and easy to use.
This bundle of sensors supports experiments in chemistry.
Ordering information: Required but not included: NeuLog USB module or NeuLog bluetooth module (which also provide power for the sensors), or a NeuLog WiFi module plus a NeuLog rechargeable battery.
Delivery information: Bundle includes one each of the following sensors: Voltage, wide range temperature, pH, pressure, and conductivity.
Caution: For educational use only.
Spécifications
  • Désignation:
    NeuLog chemistry bundle