Neulog Biology Bundle | Avantor
Neulog Biology Bundle
NeuLog biology bundle
Catalog # 470117-614
Supplier:   Avantor
Neulog Biology Bundle
Catalog # 470117-614
Supplier:  Avantor
Neulog Biology Bundle
Neulog Biology Bundle
NeuLog biology bundle
Price
$2,015.85
Log in pour la disponibilité
Pack of 1
Product Details & Documents
A set of six NeuLog sensors that are great for use in biology classrooms.
  • Collect data in the classroom or remotely
  • Compatible with PC, Mac, and chromebook
  • Free software available for download
  • Connect multiple sensors simultaneously to a single battery or connectivity module
The NeuLog probeware system is a complete data collection platform perfect for classroom use. Data are sent from the sensors to any computer via USB, bluetooth, or WiFi. A rechargeable battery makes remote data collection easy: the sensors will store the data for pre-defined experiments until they are uploaded to a computer later. The free software is powerful and easy to use.
This bundle of sensors supports experiments in biology.
Ordering information: Required but not included: NeuLog USB module or NeuLog bluetooth module (which also provide power for the sensors), or a NeuLog WiFi module plus a NeuLog rechargeable battery.
Delivery information: Bundle includes one each of the following sensors: CO₂ gas, oxygen gas, temperature, pressure, conductivity, and heart rate/pulse.
Caution: For educational use only.
Spécifications
  • Désignation:
    NeuLog biology bundle