PC Starter Neulog
Neulog PC starter bundle
Catalog # 470117-610
Supplier: Avantor
Jump to:
Product Details & Documents
Includes a NeuLog USB module and rechargeable battery module to allow for both live data collection on computers and off line data collection with sensors powered by the battery. Sensors not included.
Spécifications
- Désignation:Neulog PC starter bundle