NeuLog WiFi Bundle
NeuLog WiFi bundle
Catalog # 470117-622
Supplier: Avantor
Jump to:
Product Details & Documents
Connect any of the NeuLog sensors to your WiFi device.
- Collect data in the classroom or remotely
- Compatible with PC, Mac, and chromebook
- Free software available for download
- Connect multiple sensors simultaneously
Connect any of the NeuLog sensors to your WiFi device. The NeuLog WiFi module can be connected to computers with included USB cable or used with the included rechargeable battery to connect to any WiFi device.
Ordering information: Sensors not included.
Delivery information: Bundle includes: (1) NeuLog WiFi module and (1) rechargeable battery module.
Caution: For educational use only.
Spécifications
- Désignation:NeuLog WiFi bundle
- Cat. No.:470117-622
- Interface:WiFi