ContactUsLinkComponent
Two-Stroke Diesel Engine Model
Catalogue # 470157-714
Fournisseur:  Wards
undefined
Two-Stroke Diesel Engine Model
Catalogue # 470157-714
Fournisseur:  Wards
Supplier Number:  PH0490
Spécifications
  • Description:
    Two-Stroke Diesel Engine Model
  • Cat. no.:
    470157-714
  • Supplier No.:
    PH0490
Spécifications
À propos de cet article
This model shows the internal structure and operating principles of an air cooled two-stroke engine. All parts are aluminum and plastic. Ignition is shown via a small bulb. Carburetor and fuel supply are also sectioned. Hand crank is provided for manual operation. Mounted on a base.